ENA388699601 Enamine

6-(aminooxy)hexanoic acid hydrochloride

  • MDL number MFCD23144036
  • Cas Number : 511545-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122962
Availability details for
Quantity
  • Availability
  • Add to Cart