ENA388699613 Enamine

1-(2,2-difluoroethyl)-1,4-diazepane hydrochloride

  • MDL number MFCD23144050
  • Cas Number : 1427380-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123089
Availability details for
Quantity
  • Availability
  • Add to Cart