ENA388699614 Enamine

(4-chloro-3-fluorophenyl)(5-methyl-1,2,4-oxadiazol-3-yl)methanamine hydrochloride

  • MDL number MFCD23144051
  • Cas Number : 1427379-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123100

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123100
Availability details for
Quantity
  • Availability
  • Add to Cart