ENA388699618 Enamine

tert-butyl 3-[(3-chlorophenyl)methyl]-3-hydroxypyrrolidine-1-carboxylate

  • MDL number MFCD23144059
  • Cas Number : 1427380-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123152

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123152
Availability details for
Quantity
  • Availability
  • Add to Cart