ENA388699619 Enamine

8-oxa-1-azaspiro[4.5]decane hydrochloride

  • MDL number MFCD23144063
  • Cas Number : 1427378-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123162
Availability details for
Quantity
  • Availability
  • Add to Cart