ENA388699626 Enamine

2-(1-methyl-3-phenyl-1H-1,2,4-triazol-5-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD23144075
  • Cas Number : 1427378-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123304

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123304
Availability details for
Quantity
  • Availability
  • Add to Cart