ENA388699634 Enamine

(3,4-difluorophenyl)(1,2,4-oxadiazol-3-yl)methanamine hydrochloride

  • MDL number MFCD23144087
  • Cas Number : 1427380-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123385

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123385
Availability details for
Quantity
  • Availability
  • Add to Cart