ENA388699637 Enamine

4-(aminomethyl)cyclohexan-1-amine dihydrochloride

  • MDL number MFCD23144092
  • Cas Number : 1427380-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-123420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123420
Availability details for
Quantity
  • Availability
  • Add to Cart