ENA388699639 Enamine

methyl 3-(3-amino-2-oxopyrrolidin-1-yl)propanoate hydrochloride

  • MDL number MFCD23144094
  • Cas Number : 1427378-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123453
Availability details for
Quantity
  • Availability
  • Add to Cart