ENA388699640 Enamine

1-(2-phenylethyl)-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine dihydrochloride

  • MDL number MFCD23144095
  • Cas Number : 1427380-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 234...236
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123455

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123455
Availability details for
Quantity
  • Availability
  • Add to Cart