ENA388699642 Enamine

4-amino-5-(4-chlorophenyl)-1-methylpyrrolidin-2-one hydrochloride

  • MDL number MFCD23144097
  • Cas Number : 1427379-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123459

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123459
Availability details for
Quantity
  • Availability
  • Add to Cart