ENA388699644 Enamine

2-amino-N-hydroxy-2-methylpropanamide hydrochloride

  • MDL number MFCD23144100
  • Cas Number : 26815-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181...183
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123467

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123467
Availability details for
Quantity
  • Availability
  • Add to Cart