ENA388699648 Enamine

(9H-fluoren-9-yl)methyl N-(5,5-dimethyl-2-oxooxolan-3-yl)carbamate

  • MDL number MFCD23144105
  • Cas Number : 1427378-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123509
Availability details for
Quantity
  • Availability
  • Add to Cart