ENA388699655 Enamine

4-propyl-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD23144113
  • Cas Number : 1427380-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123575

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123575
Availability details for
Quantity
  • Availability
  • Add to Cart