ENA388699657 Enamine

2-{4-[(tert-butoxy)carbonyl]piperazin-1-yl}-2-methylpropanoic acid hydrochloride

  • MDL number MFCD23144115
  • Cas Number : 1427380-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123582
Availability details for
Quantity
  • Availability
  • Add to Cart