ENA388699659 Enamine

3,3-dimethyl-4-(methylamino)butan-1-ol hydrochloride

  • MDL number MFCD23144117
  • Cas Number : 1427379-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123598
Availability details for
Quantity
  • Availability
  • Add to Cart