ENA388699662 Enamine

3-ethyl-5-methoxy-1-methyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD23144124
  • Cas Number : 1427380-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-123749
Availability details for
Quantity
  • Availability
  • Add to Cart