ENA388699663 Enamine

1-(2-aminoethyl)-3-methyl-1,2-dihydroquinoxalin-2-one hydrochloride

  • MDL number MFCD23144125
  • Cas Number : 1427380-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123754

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123754
Availability details for
Quantity
  • Availability
  • Add to Cart