ENA388699666 Enamine

1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethane-1,2-diamine dihydrochloride

  • MDL number MFCD23144129
  • Cas Number : 1427380-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123806

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123806
Availability details for
Quantity
  • Availability
  • Add to Cart