ENA388699679 Enamine

2-methyl-2-(pyridin-3-yl)propanoic acid hydrochloride

  • MDL number MFCD23144144
  • Cas Number : 1427380-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123874

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123874
Availability details for
Quantity
  • Availability
  • Add to Cart