ENA388699681 Enamine

4-amino-N-tert-butylpiperidine-1-carboxamide dihydrochloride

  • MDL number MFCD23144146
  • Cas Number : 1427379-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-123893

Safety Information

msds https://www.enaminestore.com/catalog/EN300-123893
Availability details for
Quantity
  • Availability
  • Add to Cart