ENA388699692 Enamine

(1-methyl-1H-1,2,4-triazol-3-yl)methanamine hydrochloride

  • MDL number MFCD23144165
  • Cas Number : 215871-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124062

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124062
Availability details for
Quantity
  • Availability
  • Add to Cart