ENA388699694 Enamine

ethyl 2-[(2-carbamoylphenyl)amino]acetate

  • MDL number MFCD23144167
  • Cas Number : 105234-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124077
Availability details for
Quantity
  • Availability
  • Add to Cart