ENA388699695 Enamine

(1-phenyl-1H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD22055870
  • Cas Number : 2518-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124117

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-124117
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart