ENA388699696 Enamine

ethyl 1-(piperidin-3-yl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD23144168
  • Cas Number : 1427380-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124130
Availability details for
Quantity
  • Availability
  • Add to Cart