ENA388699702 Enamine

2-(aminomethyl)-1,4lambda6-oxathiane-4,4-dione hydrochloride

  • MDL number MFCD23144178
  • Cas Number : 1427379-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124213
Availability details for
Quantity
  • Availability
  • Add to Cart