ENA388699714 Enamine

1-{[3-(4-methylphenyl)-1,2-oxazol-5-yl]methyl}pyrrolidin-3-amine dihydrochloride

  • MDL number MFCD23144193
  • Cas Number : 1427378-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124344
Availability details for
Quantity
  • Availability
  • Add to Cart