ENA388699716 Enamine

4-(3-bromophenyl)-4-methylpiperidine hydrochloride

  • MDL number MFCD23144195
  • Cas Number : 1427380-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124355
Availability details for
Quantity
  • Availability
  • Add to Cart