ENA388699720 Enamine

1-(3-aminopropyl)-3-methyl-1,2-dihydroquinoxalin-2-one dihydrochloride

  • MDL number MFCD23144201
  • Cas Number : 1427378-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124390

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124390
Availability details for
Quantity
  • Availability
  • Add to Cart