ENA388699722 Enamine

ethyl 5-bromo-2,3-dihydro-1-benzofuran-3-carboxylate

  • MDL number MFCD23144204
  • Cas Number : 93670-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124431
Availability details for
Quantity
  • Availability
  • Add to Cart