ENA388699724 Enamine

5-[(dimethylamino)methyl]-1,2-oxazole-3-carboxylic acid hydrochloride

  • MDL number MFCD23144207
  • Cas Number : 1427378-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-124457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124457
Availability details for
Quantity
  • Availability
  • Add to Cart