ENA388699735 Enamine

2-[(2-chlorophenyl)sulfanyl]acetohydrazide hydrochloride

  • MDL number MFCD23144218
  • Cas Number : 1427380-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124489

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124489
Availability details for
Quantity
  • Availability
  • Add to Cart