ENA388699738 Enamine

ethyl 2-[4-(aminomethyl)-3,5-dimethyl-1H-pyrazol-1-yl]acetate hydrochloride

  • MDL number MFCD23144221
  • Cas Number : 1427379-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124561

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124561
Availability details for
Quantity
  • Availability
  • Add to Cart