ENA388699741 Enamine

(2-methyl-2,3-dihydro-1H-inden-2-yl)methanamine hydrochloride

  • MDL number MFCD23144224
  • Cas Number : 129766-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124569
Availability details for
Quantity
  • Availability
  • Add to Cart