ENA388699747 Enamine

1-({2-methyl-5H,6H,7H,8H-imidazo[1,2-a]pyridin-3-yl}methyl)piperazine

  • MDL number MFCD23143985
  • Cas Number : 1427380-25-3
  • Molecular Weight : 234.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-115565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115565
Availability details for
Quantity
  • Availability
  • Add to Cart