ENA388699753 Enamine

2-(3-fluorobenzoyl)pyridine

  • MDL number MFCD07699232
  • Cas Number : 898779-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37...39
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122932

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122932
Availability details for
Quantity
  • Availability
  • Add to Cart