ENA388699758 Enamine

5-amino-6-chloropyridine-3-carboxylic acid

  • MDL number MFCD18259039
  • Cas Number : 72587-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122937

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122937
Availability details for
Quantity
  • Availability
  • Add to Cart