ENA388699761 Enamine

3,4-diamino-N,N-dimethylbenzamide

  • MDL number MFCD18338463
  • Cas Number : 345205-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122940
Availability details for
Quantity
  • Availability
  • Add to Cart