ENA388699762 Enamine

1-amino-4-fluoro-2,3-dihydro-1H-indene-1-carboxylic acid

  • MDL number MFCD18678278
  • Cas Number : 1270435-39-6
  • Molecular Weight : 195.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122942
Availability details for
Quantity
  • Availability
  • Add to Cart