ENA388699771 Enamine

2-cyclohexyl-2-methylpropan-1-amine

  • MDL number MFCD20699960
  • Cas Number : 143689-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122952

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
hcodes C,Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-122952
pictograms GHS05,GHS07
hstatements H302,H312,H314,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart