ENA388699772 Enamine

5-propyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD09738121
  • Cas Number : 131609-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122953

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122953
Availability details for
Quantity
  • Availability
  • Add to Cart