ENA388699775 Enamine

2-[3-(benzyloxy)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD01657170
  • Cas Number : 29973-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122956

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122956
Availability details for
Quantity
  • Availability
  • Add to Cart