ENA388699783 Enamine

2-[4-(difluoromethoxy)phenyl]morpholine

  • MDL number MFCD11646247
  • Cas Number : 1097820-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122964
Availability details for
Quantity
  • Availability
  • Add to Cart