ENA388699785 Enamine

N-(prop-2-yn-1-yl)propanamide

  • MDL number MFCD14631308
  • Cas Number : 18327-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122966
Availability details for
Quantity
  • Availability
  • Add to Cart