ENA388699788 Enamine

2-[(2-hydroxyphenyl)sulfanyl]acetic acid

  • MDL number MFCD16159876
  • Cas Number : 785793-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122969

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122969
Availability details for
Quantity
  • Availability
  • Add to Cart