ENA388699791 Enamine

4-({[(oxan-2-yl)methyl](propan-2-yl)amino}methyl)aniline

  • MDL number MFCD12587186
  • Cas Number : 1154967-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122972
Availability details for
Quantity
  • Availability
  • Add to Cart