ENA388699793 Enamine

1-benzyl-2,5-dimethylpiperazine

  • MDL number MFCD12761573
  • Cas Number : 29906-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115844
Availability details for
Quantity
  • Availability
  • Add to Cart