ENA388699796 Enamine

6-phenyl-1-benzofuran

  • MDL number MFCD18451259
  • Cas Number : 35664-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122989
Availability details for
Quantity
  • Availability
  • Add to Cart