ENA388699817 Enamine

1-phenyl-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine

  • MDL number MFCD20429999
  • Cas Number : 87673-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116793
Availability details for
Quantity
  • Availability
  • Add to Cart