ENA388699821 Enamine

3-[4-(propan-2-yl)piperazin-1-yl]aniline

  • MDL number MFCD10032984
  • Cas Number : 1018648-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116970
Availability details for
Quantity
  • Availability
  • Add to Cart